missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) carbonate, Mn 44% min

For Pigment, Varnish Drier. | CAS: 598-62-9 | CMnO3 | 114.946 g/mol

$102.22 - $257.74

Chemical Identifiers

CAS 598-62-9
Molecular Formula CMnO3
Molecular Weight (g/mol) 114.946
MDL Number MFCD00011116
InChI Key XMWCXZJXESXBBY-UHFFFAOYSA-L
Synonym manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
PubChem CID 11726
IUPAC Name manganese(2+);carbonate
SMILES C(=O)([O-])[O-].[Mn+2]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1432436
View Documents
Thermo Scientific Chemicals
01432436
500 g
Each for $102.22
Only null left
 
AA14324A3
View Documents
Thermo Scientific Chemicals
014324A3
2 kg
Each for $257.74
Only null left
 
Description

Description

Pigment, varnish drierManganese(II) carbonate is used as an additive to plant fertilizers, glaze colorant in ceramics, pigment and drier for varnishes. It is also used as an ingredient in welding rod, animal feed additive as well as to prepare other manganese salts and pharmaceuticals. It is also used in medicine as a hematinic and in health foods, in flux and in concrete stains.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

598-62-9
114.946
XMWCXZJXESXBBY-UHFFFAOYSA-L
11726
C(=O)([O-])[O-].[Mn+2]
CMnO3
MFCD00011116
manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
manganese(2+);carbonate

Specifications

350°C (decomposition)
598-62-9
500 g
CMnO3
manganese carbonate, manganese ii carbonate, manganous carbonate, manganese 2+ carbonate, natural rhodochrosite, manganese carbonate 1:1, rhodochrosite, manganese carbonate mnco3, unii-9zv57512zm, ccris 3660
XMWCXZJXESXBBY-UHFFFAOYSA-L
manganese(2+);carbonate
11726
Odorless
3.125
Brown to Violet
Powder
Mn 44% min.
MFCD00011116
Soluble in dilute inorganic acids and carbon dioxide. Insoluble in water,alcohol,ammonia and common organic acids.
C(=O)([O-])[O-].[Mn+2]
114.946
114.95
Light brown to violet powder
Manganese(II) carbonate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 209-942-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only