missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) bromide hydrate, 99.9% (metals basis)

Catalog No. p-7059343
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4444309 10 g
AA4444318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4444309 Supplier Thermo Scientific Chemicals Supplier No. 04444309
Only null left

CAS: 10031-20-6 | Br2H8MnO4 | 286.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10031-20-6
Molecular Formula Br2H8MnO4
Molecular Weight (g/mol) 286.81
MDL Number MFCD00149791
InChI Key HHDPJRSXPOGIOP-UHFFFAOYSA-L
Synonym manganese bromide tetrahydrate, unii-8308v6rbj5, manganese ii bromide hexahydrate, manganous bromide tetrahydrate, manganese dibromide tetrahydrate, manganese bromide mnbr2 , tetrahydrate, manganese 2+ ion tetrahydrate dibromide
IUPAC Name manganese(2+) tetrahydrate dibromide
SMILES O.O.O.O.[Mn++].[Br-].[Br-]

Specifications

Melting Point 64.3°C (decomposition)
Color Pink
Physical Form Crystalline
Quantity 10 g
Merck Index 14,5725
Formula Weight 214.76 (Anhydrous)
Percent Purity 99.9%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Manganese (II) bromide hydrate
Show More Show Less
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 236-591-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only