missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) bromide, anhydrous, 95%

Catalog No. p-7024113
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1832114 25 g
AAA1832122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1832114 Supplier Thermo Scientific Chemicals Supplier No. A1832114
Only null left

CAS: 13446-03-2 | MnBr2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13446-03-2
Molecular Formula MnBr2
MDL Number MFCD00011113

Specifications

Melting Point 698°C
Quantity 25 g
Merck Index 14,5725
Formula Weight 214.76
Percent Purity 95%
Odor Odorless
Sensitivity Hygroscopic
Density 4.38
Chemical Name or Material Manganese(II) bromide, anhydrous
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 236-591-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only