missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) bromide, anhydrous, 95%

Catalog No. AAA1832122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1832122 100 g
AAA1832114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1832122 Supplier Thermo Scientific Chemicals Supplier No. A1832122

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 13446-03-2 | MnBr2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13446-03-2
Molecular Formula MnBr2
MDL Number MFCD00011113

Specifications

Melting Point 698°C
Quantity 100 g
Odor Odorless
Sensitivity Hygroscopic
Density 4.38
Merck Index 14,5725
Formula Weight 214.76
Percent Purity 95%
Chemical Name or Material Manganese(II) bromide, anhydrous
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 236-591-9
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only