missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™

Catalog No. M28621G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28621G 1 g
M28625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28621G Supplier TCI America Supplier No. M28621G

Chemical Identifiers

CAS 207861-55-0
Molecular Formula C4F12MnN2O8S4
Molecular Weight (g/mol) 615.209
MDL Number MFCD23380173
InChI Key NNWDGVNRIAAYMY-UHFFFAOYSA-N
Synonym manganese bis-trifluoromethanesulfonimide
PubChem CID 134159301
IUPAC Name bis(trifluoromethylsulfonyl)azanide;manganese(2+)
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Mn+2]

Specifications

Color White-Yellow
Quantity 1 g
UN Number 1759
Formula Weight 615.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Manganese(II) Bis(trifluoromethanesulfonyl)imide