missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) acetate tetrahydrate, Mn 22% (typical)

Catalog No. p-7048983
Change view
Click to view available options
Quantity:
100 g
500 g
2 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1235122 100 g
AA1235136 500 g
AA12351A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1235122 Supplier Thermo Scientific Chemicals Supplier No. 01235122
Only null left

CAS: 6156-78-1 | C4H14MnO8 | 245.09 g/mol

It is used as a mordant in dyeing, drier for paints and varnishes, an oxidation catalyst, and in the manufacture of bister. It is also used in the manufacture of fertilizer. It is used for the production of high purity manganese dioxide, manganese oxide hydrate, oxide-based catalysts, and catalysts for oxidation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Mordant in dyeing, in the manufacture of bister, as a drier for paints and varnishes, and as an oxidation catalyst

Chemical Identifiers

CAS 6156-78-1
Molecular Formula C4H14MnO8
Molecular Weight (g/mol) 245.09
MDL Number MFCD00062552
InChI Key CESXSDZNZGSWSP-UHFFFAOYSA-L
Synonym manganese ii acetate tetrahydrate, manganese acetate tetrahydrate, manganous acetate tetrahydrate, unii-9to51d176n, manganese diacetate, tetrahydrate, manganese 2+ diacetate tetrahydrate, acetic acid, manganese 2+ salt, tetrahydrate, manganese ii acetatetetrahydrate, acmc-20akkp
PubChem CID 93021
IUPAC Name manganese(2+);diacetate;tetrahydrate
SMILES O.O.O.O.[Mn++].CC([O-])=O.CC([O-])=O

Specifications

Melting Point 80°C
Color Pink
Density 1.589 g/mL
Odor Pungent
Linear Formula Mn(OOCCH3)2·4H2O
Quantity 100 g
Merck Index 14,5724
Solubility Information Soluble in water,methanol,ethanol and acetic acid.
Formula Weight 245.09 (173.03 Anhydrous)
Concentration or Composition (by Analyte or Components) Mn 22% (typical)
Physical Form Crystalline
Chemical Name or Material Manganese(II) acetate tetrahydrate
Show More Show Less
Hazard Category H315-H319-H335-H336-H373
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c
EINECSNumber 211-334-3
RTECSNumber AI5775000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only