missing translation for 'onlineSavingsMsg'
Learn More

Manganese(II) acetate, 98+%, anhydrous

Catalog No. p-3523483
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC389140050 5 g Glass bottle
AC389140250 25 g Glass bottle
AC389141000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC389140050 Supplier Thermo Scientific Chemicals Supplier No. 389140050
Only null left

CAS: 638-38-0 | C4H6MnO4 | 173.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 638-38-0
Molecular Formula C4H6MnO4
Molecular Weight (g/mol) 173.03
MDL Number MFCD00013039
InChI Key UOGMEBQRZBEZQT-UHFFFAOYSA-L
Synonym manganese ii acetate, manganese acetate, manganous acetate, diacetylmanganese, manganese diacetate, manganese di acetate, manganese 2+ acetate, octan manganaty czech, acetic acid, manganese 2+ salt, unii-0v6e9q2i0y
PubChem CID 12525
IUPAC Name manganese(2+);diacetate
SMILES [Mn++].CC([O-])=O.CC([O-])=O

Specifications

Color Beige-Gray to Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 98% min. (Complexometry)
Linear Formula Mn(OC(O)CH3)2
Merck Index 15, 5789
Solubility Information Solubility in water: very soluble
Formula Weight 173.03
Percent Purity ≥98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Manganese(II) acetate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Con
EINECSNumber 211-334-3

RUO – Research Use Only