missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese ICP Standard, 10,000 ppm Mn in 5% HNO3, Ricca Chemical
Specifications
Specifications
| Packaging | Natural Poly Bottle |
| CAS | 6156-78-1, 7697-37-2, 7732-18-5 |
| Concentration or Composition Notes | 10,000ppm Mn |
| CAS Min % | 88.0, 6.58, 3.91 |
| CAS Max % | 91.0, 6.84, 4.07 |
| Equivalence | 1 mL = 10 mg Mn |
| Molecular Formula | C4H14MnO8 |
| InChI Key | CESXSDZNZGSWSP-UHFFFAOYSA-L |
| SMILES | CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Mn+2] |
| IUPAC Name | manganese(2+);diacetate;tetrahydrate |
| Show More |