missing translation for 'onlineSavingsMsg'
Learn More

Manganese ICP Standard, 10,000 ppm Mn in 5% HNO3, Ricca Chemical

Supplier:  Ricca Chemical Company PMN10KN100

Catalog No. PMN10KN100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Manganese ICP Standard
6156-78-1 , 7697-37-2 , 7732-18-5
88.0, 6.58, 3.91
1 mL = 10 mg Mn
CESXSDZNZGSWSP-UHFFFAOYSA-L
manganese(2+);diacetate;tetrahydrate
93021
Natural LDPE Bottle
10,000ppm Mn
91.0, 6.84, 4.07
C4H14MnO8
CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Mn+2]
245.086
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents