missing translation for 'onlineSavingsMsg'
Learn More

Manganese Glycerophosphate, Powder, FCC, 98-100.5%, Spectrum™ Chemical

Catalog No. 18604200
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604200 500 g Poly Bottle
18604201 2.5 kg Poly Pail
18604202 12 kg Poly Pail
18604203 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18604200 Supplier Spectrum Chemical Mfg Cor Supplier No. MA163500GM
Only null left

MA163, 1320-46-3, C3H7MnO6P·xH2O

Spectrum™ Chemical Manganese Glycerophosphate, Powder, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1320-46-3
Molecular Formula C8H18Mn2O12P2
Molecular Weight (g/mol) 478.04
InChI Key PNNHLULHRSZWPG-UHFFFAOYNA-J
IUPAC Name dimanganese(2+) 3-(phosphonatooxy)pentane-1,5-diol 3-(phosphonatooxy)propane-1,2-diol
SMILES [Mn++].[Mn++].OCC(O)COP([O-])([O-])=O.OCCC(CCO)OP([O-])([O-])=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98 to 100.5%
Grade FCC
Loss on Drying 12%
Packaging Poly Bottle