missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese Gluconate, Dihydrate, USP, 98-102%, Spectrum™ Chemical
Description
Spectrum™ Chemical Manganese Gluconate, Dihydrate, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Quantity | 500 g |
| Percent Purity | 98 to 102% |
| Grade | USP |
| Packaging | Poly Bottle |