missing translation for 'onlineSavingsMsg'
Learn More

Manganese Dioxide, Powder, Technical, Spectrum™ Chemical

Catalog No. p-7165831
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611490 500 g Amber Glass Bottle
18611492 2.5 kg Amber Glass Bottle
18611489 12 kg Poly Pail
18611491 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611490 Supplier Spectrum Chemical Mfg Cor Supplier No. M1108500GM
Only null left

Chemical Identifiers

CAS 1313-13-9

Specifications

CAS Max % 1
Quantity 500 g
Grade Technical
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.