missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese Chloride, Tetrahydrate, USP, 98-101%, Spectrum™ Chemical
M1106, 13446-34-9, MnCl2·4H2O
Supplier: Spectrum Chemical Mfg Cor M1106500GM
Description
Spectrum™ Chemical Manganese Chloride, Tetrahydrate, USP is indicated for use as a supplement to intravenous solutions given for total parenteral nutrition (TPN) to maintain Manganese serum levels. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 100% | |
| Cl2H8MnO4 | |
| CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
| manganese(2+) tetrahydrate dichloride | |
| 98 to 101% | |
| 36.0-38.5% |
| 500 g | |
| MFCD00149792 | |
| O.O.O.O.[Cl-].[Cl-].[Mn++] | |
| 197.90 | |
| USP | |
| Poly Bottle |
Chemical Identifiers
| Cl2H8MnO4 | |
| MFCD00149792 | |
| manganese(2+) tetrahydrate dichloride |
| 197.90 | |
| CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
| O.O.O.O.[Cl-].[Cl-].[Mn++] |