missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride, Tetrahydrate, USP, 98-101%, Spectrum™ Chemical

Catalog No. 18604183
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604183 125 g Poly Bottle
18604184 500 g Poly Bottle
18604185 2.5 kg Poly Bottle
18604186 12 kg Poly Pail
18604187 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604183 Supplier Spectrum Chemical Mfg Cor Supplier No. M1106125GM
Get Up to $500 in Chemicals with Purchase
View all promotions

M1106, 13446-34-9, MnCl2·4H2O

Spectrum™ Chemical Manganese Chloride, Tetrahydrate, USP is indicated for use as a supplement to intravenous solutions given for total parenteral nutrition (TPN) to maintain Manganese serum levels. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

CAS Max % 100%
Quantity 125 g
Percent Purity 98 to 101%
Grade USP
Loss on Drying 36.0-38.5%
Packaging Poly Bottle