missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese Chloride, Tetrahydrate, FCC, 98-102%, Spectrum™ Chemical
M1109, 13446-34-9, MnCl2·4H2O
$438.00 - $18693.79
Chemical Identifiers
| Molecular Formula | Cl2H8MnO4 |
|---|---|
| Molecular Weight (g/mol) | 197.90 |
| MDL Number | MFCD00149792 |
| InChI Key | CNFDGXZLMLFIJV-UHFFFAOYSA-L |
| SMILES | O.O.O.O.[Cl-].[Cl-].[Mn++] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604178
|
Spectrum Chemical Mfg Cor
M1109125GM |
125 g | Poly Bottle |
Each for $438.00
|
|
||||
|
18604179
|
Spectrum Chemical Mfg Cor
M1109500GM |
500 g | Poly Bottle |
Each for $1,070.15
|
|
||||
|
18604180
|
Spectrum Chemical Mfg Cor
M110925KG |
2.5 kg | Poly Pail |
Each for $2,765.58
|
|
||||
|
18604182
|
Spectrum Chemical Mfg Cor
M110925KGBL |
25 kg | Fiber Drum |
Each for $9,589.76
|
|
||||
|
18604181
|
Spectrum Chemical Mfg Cor
M110950KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Manganese Chloride, Tetrahydrate, FCC is used as a dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| Cl2H8MnO4 | |
| MFCD00149792 | |
| O.O.O.O.[Cl-].[Cl-].[Mn++] |
| 197.90 | |
| CNFDGXZLMLFIJV-UHFFFAOYSA-L |
Specifications
| 13446-34-9 | |
| 125 g | |
| MFCD00149792 | |
| O.O.O.O.[Cl-].[Cl-].[Mn++] | |
| 98 to 102% | |
| Poly Bottle |
| 100% | |
| Cl2H8MnO4 | |
| CNFDGXZLMLFIJV-UHFFFAOYSA-L | |
| 197.90 | |
| FCC |