missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride Tetrahydrate, (Crystalline),MP Biomedicals

Catalog No. ICN15533490
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15533490 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15533490 Supplier MP Biomedicals Inc Supplier No. 0215533490

missing translation for 'validMainframeMsg'

High-purity crystalline manganese chloride used as a manganese ion source in various biological research applications.

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

Physical Form Crystalline
Quantity 500 g
Formula Weight 197.9
Grade Research
Chemical Name or Material Manganese Chloride Tetrahydrate
Recommended Storage Room Temperature