missing translation for 'onlineSavingsMsg'
Learn More

Manganese Chloride, Tetrahydrate, Crystal, ACS, 98-101%, Spectrum™ Chemical

Catalog No. p-7165829
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611484 125 g Amber Glass Bottle
18611485 500 g Amber Glass Bottle
18611486 2.5 kg Amber Glass Bottle
18611487 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18611484 Supplier Spectrum Chemical Mfg Cor Supplier No. M1105125GM

Chemical Identifiers

CAS 13446-34-9
Molecular Formula Cl2H8MnO4
Molecular Weight (g/mol) 197.90
MDL Number MFCD00149792
InChI Key CNFDGXZLMLFIJV-UHFFFAOYSA-L
IUPAC Name manganese(2+) tetrahydrate dichloride
SMILES O.O.O.O.[Cl-].[Cl-].[Mn++]

Specifications

CAS Max % 1
pH 3.5 to 6.0
Quantity 125 g
Percent Purity 98 to 101%
Grade ACS
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.