missing translation for 'onlineSavingsMsg'
Learn More

(±)-Mandelic Acid, 99%

Catalog No. p-3344278
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
2500 g
500 g
Catalog No. Quantity
AAA1422322 100 g
AAA1422336 500 g
AAA142230E 2500 g
3 options

Catalog No. AAA1422322

Supplier: Thermo Scientific Chemicals A1422322

Only null left

CAS: 90-64-2 | C8H8O3 | 152.149 g/mol

Mandelic acid is used in the pharmaceutical industry as an antibacterial to treat urinary tract infections. It acts as a precursor for making various drugs. It is also used as an oral antibiotic. Mandelic acid peels are widely used for treating damaged skin due to its less penetrating and irritating property. Enantiomeric resolution of (±)-Mandelic acid is performed by using the chiral amine (1R,2S)-(-)-ephedrine followed by acidification to prepare (R)-( -)-Mandelic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mandelic acid is used in the pharmaceutical industry as an antibacterial to treat urinary tract infections. It acts as a precursor for making various drugs. It is also used as an oral antibiotic. Mandelic acid peels are widely used for treating damaged skin due to its less penetrating and irritating property. Enantiomeric resolution of (±)-Mandelic acid is performed by using the chiral amine (1R,2S)-(-)-ephedrine followed by acidification to prepare (R)-( -)-Mandelic acid.

Solubility
Soluble in water, diethyl ether, ethanol and isopropanol.

Notes
Hydrophobic and light sensitive. Incompatible with strong oxidizing agents and reducing agents.

Chemical Identifiers

CAS 90-64-2
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00064250
InChI Key IWYDHOAUDWTVEP-UHFFFAOYSA-N
Synonym mandelic acid, dl-mandelic acid, phenylglycolic acid, amygdalic acid, almond acid, uromaline, p-mandelic acid, racemic mandelic acid, paramandelic acid, rs-mandelic acid
PubChem CID 1292
ChEBI CHEBI:35825
IUPAC Name 2-hydroxy-2-phenylacetic acid
SMILES C1=CC=C(C=C1)C(C(=O)O)O

Specifications

Melting Point 118°C to 121°C
Assay Percent Range 99%
Beilstein 510011
Merck Index 14,5717
Solubility Information Soluble in water,diethyl ether,ethanol and isopropanol.
Formula Weight 152.15
Percent Purity 99%
Quantity 100 g
Sensitivity Light sensitive
Chemical Name or Material (±)-Mandelic Acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H318
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 202-007-6
RTECSNumber OO6300000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only