missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Maltotriose, 98%

Catalog No. AAJ6649103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6649103 1 g
AAJ6649106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6649103 Supplier Thermo Scientific Chemicals Supplier No. J6649103

CAS: 1109-28-0 | C18H32O16 | 504.438 g/mol

Maltotriose is a matrose regulon inducer in Escherichia coli. Maltotriose has been used in a study to investigate the influence of fourth generation poly(propyleneimine) dendrimers on blood cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maltotriose is a matrose regulon inducer in Escherichia coli. Maltotriose has been used in a study to investigate the influence of fourth generation poly(propyleneimine) dendrimers on blood cells.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1109-28-0
Molecular Formula C18H32O16
Molecular Weight (g/mol) 504.438
MDL Number MFCD00006629
InChI Key FYGDTMLNYKFZSV-NBCPLHMPSA-N
Synonym maltotriose, d-maltotriose
PubChem CID 134129496
IUPAC Name (2R,3S,4S,5S,6R)-2-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4S,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O

Specifications

Melting Point 132°C to 135°C
Color White
Quantity 1 g
Beilstein 1443481
Solubility Information Soluble in water.
Optical Rotation +165° (c=1 in Water)
Formula Weight 504.44
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Maltotriose
EINECSNumber 214-174-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only