missing translation for 'onlineSavingsMsg'
Learn More

Maltotriose, 93%

CAS: 1109-28-0 | C18H32O16 | 504.44 g/mol

Supplier:  Thermo Scientific Chemicals 225940250

 View more versions of this product

Catalog No. AC225940250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Maltotriose
132.0°C to 135.0°C
Authentic
Glass bottle
MFCD00006629
+162.50° (25°C c=1,H2O)
maltotriose, d-maltotriose
FYGDTMLNYKFZSV-NBCPLHMPSA-N
(2R,3S,4S,5S,6R)-2-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4S,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
134129496
93%
1109-28-0
White
91% min. (ELSD) (HPLC)
C18H32O16
25 g
+ 162.50
Solubility in water: 50mg/mL,clear colorless solution
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O
504.44
504.44
Fine Crystalline Powder or Crystals

Chemical Identifiers

1109-28-0
FYGDTMLNYKFZSV-NBCPLHMPSA-N
134129496
C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O
MFCD00006629
maltotriose, d-maltotriose
(2R,3S,4S,5S,6R)-2-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4S,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 214-174-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only