missing translation for 'onlineSavingsMsg'
Learn More

Maltotriose, 93%

Catalog No. AC225945000
Change view
Click to view available options
Quantity:
500 mg
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC225945000 500 mg
AC225940250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC225945000 Supplier Thermo Scientific Chemicals Supplier No. 225945000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 1109-28-0 | C18H32O16 | 504.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1109-28-0
MDL Number MFCD00006629
InChI Key FYGDTMLNYKFZSV-NBCPLHMPSA-N
Synonym maltotriose, d-maltotriose
PubChem CID 134129496
IUPAC Name (2R,3S,4S,5S,6R)-2-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-2-(hydroxymethyl)-6-[(2R,3S,4S,5R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)OC3C(OC(C(C3O)O)O)CO)CO)O)O)O)O

Specifications

Melting Point 132.0°C to 135.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 91% min. (ELSD) (HPLC)
Packaging Glass bottle
Molecular Formula C18H32O16
Quantity 500 mg
Specific Rotation Condition +162.50° (25°C c=1,H2O)
Specific Rotation + 162.50
Solubility Information Solubility in water: 50mg/mL,clear colorless solution
Molecular Weight (g/mol) 504.44
Formula Weight 504.44
Percent Purity 93%
Physical Form Fine Crystalline Powder or Crystals
Chemical Name or Material Maltotriose
Show More Show Less
EINECSNumber 214-174-2

RUO – Research Use Only