missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Maltodextrin 12

Catalog No. AC466680500
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466680500 50 g
AC466682500 250 g
AC466680010 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC466680500 Supplier Thermo Scientific Chemicals Supplier No. 466680500
Only null left

CAS: 9050-36-6 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 9050-36-6
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C=O

Specifications

Melting Point 240.0°C
Quantity 50 g
Chemical Name or Material Maltodextrin 12
EINECSNumber 232-940-4

RUO – Research Use Only