missing translation for 'onlineSavingsMsg'
Learn More

Malotilate 98.0+%, TCI America™

Catalog No. M26741G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26741G 1 g
M26745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26741G Supplier TCI America Supplier No. M26741G

Chemical Identifiers

CAS 59937-28-9
Molecular Formula C12H16O4S2
Molecular Weight (g/mol) 288.376
MDL Number MFCD00867646
InChI Key YPIQVCUJEKAZCP-UHFFFAOYSA-N
Synonym Diisopropyl (1,3-Dithiol-2-ylidene)malonate, (1,3-Dithiol-2-ylidene)malonic Acid Diisopropyl Ester
PubChem CID 4006
IUPAC Name dipropan-2-yl 2-(1,3-dithiol-2-ylidene)propanedioate
SMILES CC(C)OC(=O)C(=C1SC=CS1)C(=O)OC(C)C

Specifications

Melting Point 61°C
Color Yellow
Quantity 1 g
Formula Weight 288.38
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Malotilate
RTECSNumber OO0970000
TSCA No