missing translation for 'onlineSavingsMsg'
Learn More

Malonyl-L-Carnitine-(N-Methyl-D3) Lithium Salt Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022810
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022810 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022810 Supplier Merck Emd Millipore Supplier No. 9303510MG

Specifications

CAS 1803252-71-2 (Free Acid)
Flash Point Not applicable
Molecular Formula C10D3H14NO6 · xLi+
Linear Formula C10D3H14NO6 · xLi+
Quantity 10 mg
Optical Rotation [α]/D -28°C ± 2°C = 0.1 in. H218O
Formula Weight 250.26 (Free Acid Basis)
Percent Purity ≥92% (HPLC)
Grade Analytical Standard