missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Malonyl-L-Carnitine-(N-Methyl-D3) Lithium Salt Analytical Standard, MilliporeSigma™ Supelco™
$910.81 - $910.81
Specifications
| CAS | 1803252-71-2 (Free Acid) |
|---|---|
| Flash Point | Not applicable |
| Molecular Formula | C10D3H14NO6 · xLi+ |
| Linear Formula | C10D3H14NO6 · xLi+ |
| Optical Rotation | [α]/D -28°C ± 2°C = 0.1 in. H218O |
Specifications
| 1803252-71-2 (Free Acid) | |
| C10D3H14NO6 · xLi+ | |
| [α]/D -28°C ± 2°C = 0.1 in. H218O | |
| ≥92% (HPLC) |
| Not applicable | |
| C10D3H14NO6 · xLi+ | |
| 250.26 (Free Acid Basis) | |
| Analytical Standard |