missing translation for 'onlineSavingsMsg'
Learn More

Malononitrile 98.0+%, TCI America™

Catalog No. M003325G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M003325G 25 g
M0033100G 100 g
M0033500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M003325G Supplier TCI America Supplier No. M003325G

Chemical Identifiers

CAS 109-77-3
Molecular Formula C3H2N2
Molecular Weight (g/mol) 66.063
MDL Number MFCD00001883
InChI Key CUONGYYJJVDODC-UHFFFAOYSA-N
Synonym malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano
PubChem CID 8010
ChEBI CHEBI:33186
IUPAC Name propanedinitrile
SMILES C(C#N)C#N

Specifications

Boiling Point 220°C
Quantity 25 g
UN Number 2647
Formula Weight 66.06
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material Malononitrile
EINECSNumber (2)-1511
RTECSNumber OO3150000
TSCA Yes
Recommended Storage Refrigerator