missing translation for 'onlineSavingsMsg'
Learn More

Malononitrile, 99%

Catalog No. p-154869
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC125271000 100 g Glass bottle
AC125275000 500 g Glass bottle
2 options

Catalog No. AC125271000

Supplier: Thermo Scientific Chemicals 125271000

Only null left

CAS: 109-77-3 | C3H2N2 | 66.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 109-77-3
Molecular Formula C3H2N2
Molecular Weight (g/mol) 66.06
MDL Number MFCD00001883
InChI Key CUONGYYJJVDODC-UHFFFAOYSA-N
Synonym malononitrile, dicyanomethane, cyanoacetonitrile, malonic dinitrile, methylene cyanide, dicyanmethane, malonodinitrile, malonic acid dinitrile, methylenedinitrile, methane, dicyano
PubChem CID 8010
ChEBI CHEBI:33186
IUPAC Name propanedinitrile
SMILES C(C#N)C#N

Specifications

Melting Point 30.0°C to 34.0°C
Color Brown-Yellow to White
Density 1.0400g/mL
Boiling Point 220.0°C
Flash Point 112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula NCCH2CN
Quantity 100 g
Beilstein 02,589
Fieser 04,317
Merck Index 15,5775
Specific Gravity 1.04
Solubility Information Solubility in water: 133g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and acetone
Formula Weight 66.06
Percent Purity 99%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material Malononitrile, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove
EINECSNumber 203-703-2
RTECSNumber OO3150000
TSCA TSCA

RUO – Research Use Only