missing translation for 'onlineSavingsMsg'
Learn More

Malonamide 98.0+%, TCI America™

Catalog No. M0027500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0027500G 500 g
M002725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0027500G Supplier TCI America Supplier No. M0027500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 108-13-4
Molecular Formula C3H6N2O2
Molecular Weight (g/mol) 102.093
MDL Number MFCD00008034
InChI Key WRIRWRKPLXCTFD-UHFFFAOYSA-N
Synonym malonamide, malonodiamide, malondiamide, malonyldiamide, carboxamidoacetamide, malonic acid diamide, malonic diamide, unii-qvq8cng255, qvq8cng255, methane-1,1-dicarboxamide
PubChem CID 7911
IUPAC Name propanediamide
SMILES C(C(=O)N)C(=O)N

Specifications

Melting Point 174°C
Color White
Quantity 500 g
Formula Weight 102.09
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Malonamide
EINECSNumber (2)-0915
RTECSNumber ON9125000
TSCA Yes