missing translation for 'onlineSavingsMsg'
Learn More

Malonamide, 98%

Catalog No. AAA1793936
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1793936 500 g
AAA1793922 100 g
AAA179390E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1793936 Supplier Thermo Scientific Chemicals Supplier No. A1793936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108-13-4 | C3H6N2O2 | 102.093 g/mol

Malonamide is a reagent for fluorimetric determination of reducing carbohydrates. The malonamide-based ionic liquid extractant was used in the extraction of europium(iii) and other trivalent rare earth elements from nitric acid medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Malonamide is a reagent for fluorimetric determination of reducing carbohydrates. The malonamide-based ionic liquid extractant was used in the extraction of europium(iii) and other trivalent rare earth elements from nitric acid medium.

Solubility
Soluble in HCl.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 108-13-4
Molecular Formula C3H6N2O2
Molecular Weight (g/mol) 102.093
MDL Number MFCD00008034
InChI Key WRIRWRKPLXCTFD-UHFFFAOYSA-N
Synonym malonamide, malonodiamide, malondiamide, malonyldiamide, carboxamidoacetamide, malonic acid diamide, malonic diamide, unii-qvq8cng255, qvq8cng255, methane-1,1-dicarboxamide
PubChem CID 7911
IUPAC Name propanediamide
SMILES C(C(=O)N)C(=O)N

Specifications

Melting Point 167°C to 171°C
Quantity 500 g
Beilstein 1751401
Solubility Information Soluble in HCl.
Formula Weight 102.09
Percent Purity 98%
Chemical Name or Material Malonamide
EINECSNumber 203-553-8
RTECSNumber ON9125000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only