missing translation for 'onlineSavingsMsg'
Learn More

Cadence Science™ Malleable Stainless Steel Animal Feeding Needles

Catalog No. 14825279
Change view
Click to view available options
Gauge:
15G
18G
20G
Diameter (Metric) Ball:
1.9 mm
2.3 mm
2.9 mm
Length (English):
1.5 in.
2 in.
2.8 in.
3 in.
3.1 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Gauge Diameter (Metric) Ball Length (English)
14825279 15G 2.9 mm 3 in.
0129019 20G 1.9 mm 1.5 in.
14825277 18G 2.3 mm 2 in.
14825278 18G 2.3 mm 3.1 in.
14825276 20G 1.9 mm 2.8 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 14825279 Supplier Cadence Science™ Supplier No. POP9915

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed to meet the special needs of pharmacologists, toxicologists, pathologists, and other biomedical researchers working with laboratory animals. Cadence Science™ Malleable Stainless Steel Animal Feeding Needles are sterile and disposable.

  • Type 304 stainless steel construction
  • Malleable
  • Sterile
  • Disposable
  • 100/box, 20 x 5/sterile pouch

Specifications

Product Type Animal Feeding Needle
Material Stainless Steel (304)
Disposable Single-use
For Use With (Application) Animal feeding
Gauge 15G
Sterility Sterile
Length (English) 3 in.
Diameter (Metric) Ball 2.9 mm