missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Maleimide | |
| 94°C | |
| 125°C | |
| MFCD00005494 | |
| 2923 | |
| KWLWLQRUFFGWNG-UHFFFAOYSA-L | |
| potassium silver(1+) bis(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-ide) | |
| 10935 | |
| 97.07 | |
| Crystalline Powder |
| 541-59-3 | |
| White | |
| C8H4AgKN2O4 | |
| 25 g | |
| maleimide, 1h-pyrrole-2,5-dione, 2,5-pyrroledione, maleinimide, maleic imide, 3-pyrroline-2,5-dione, 2,5-dihydro-1h-pyrrole-2,5-dione, unii-2519r1ugp8, ccris 3408, 5-hydroxy-2h-pyrrol-2-one | |
| [K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1 | |
| 339.10 | |
| CHEBI:16072 | |
| ≥98.0% (T) |
Chemical Identifiers
| 541-59-3 | |
| 339.10 | |
| KWLWLQRUFFGWNG-UHFFFAOYSA-L | |
| 10935 | |
| potassium silver(1+) bis(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-ide) |
| C8H4AgKN2O4 | |
| MFCD00005494 | |
| maleimide, 1h-pyrrole-2,5-dione, 2,5-pyrroledione, maleinimide, maleic imide, 3-pyrroline-2,5-dione, 2,5-dihydro-1h-pyrrole-2,5-dione, unii-2519r1ugp8, ccris 3408, 5-hydroxy-2h-pyrrol-2-one | |
| CHEBI:16072 | |
| [K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1 |
Safety and Handling
EINECSNumber : (5)-0122
RTECSNumber : ON4800000
TSCA : Yes