missing translation for 'onlineSavingsMsg'
Learn More

Maleimide 98.0+%, TCI America™

Catalog No. M00185G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M00185G 5 g
M001825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M00185G Supplier TCI America Supplier No. M00185G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 541-59-3
Molecular Formula C8H4AgKN2O4
Molecular Weight (g/mol) 339.10
MDL Number MFCD00005494
InChI Key KWLWLQRUFFGWNG-UHFFFAOYSA-L
Synonym maleimide, 1h-pyrrole-2,5-dione, 2,5-pyrroledione, maleinimide, maleic imide, 3-pyrroline-2,5-dione, 2,5-dihydro-1h-pyrrole-2,5-dione, unii-2519r1ugp8, ccris 3408, 5-hydroxy-2h-pyrrol-2-one
PubChem CID 10935
ChEBI CHEBI:16072
IUPAC Name potassium silver(1+) bis(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-ide)
SMILES [K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1

Specifications

Melting Point 94°C
Color White
Boiling Point 125°C
Quantity 5 g
UN Number 2923
Formula Weight 97.07
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Maleimide
EINECSNumber (5)-0122
RTECSNumber ON4800000
TSCA Yes