missing translation for 'onlineSavingsMsg'
Learn More

Maleimide, 98%

Catalog No. p-3736820
Change view
Click to view available options
Quantity:
5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC156710050 5 g Glass bottle
AC156710100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC156710050 Supplier Thermo Scientific Chemicals Supplier No. 156710050
Only null left

CAS: 541-59-3 | C8H4AgKN2O4 | 339.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 541-59-3
Molecular Formula C8H4AgKN2O4
Molecular Weight (g/mol) 339.10
MDL Number MFCD00005494
InChI Key KWLWLQRUFFGWNG-UHFFFAOYSA-L
Synonym maleimide, 1h-pyrrole-2,5-dione, 2,5-pyrroledione, maleinimide, maleic imide, 3-pyrroline-2,5-dione, 2,5-dihydro-1h-pyrrole-2,5-dione, unii-2519r1ugp8, ccris 3408, 5-hydroxy-2h-pyrrol-2-one
PubChem CID 10935
ChEBI CHEBI:16072
SMILES [K+].[Ag+].O=C1[N-]C(=O)C=C1.O=C1[N-]C(=O)C=C1

Specifications

Melting Point 90.0°C to 94.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21,399
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,ethylether
IUPAC Name pyrrole-2,5-dione
Formula Weight 97.07
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Maleimide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
Toxic if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 208-787-4

RUO – Research Use Only