missing translation for 'onlineSavingsMsg'
Learn More

Maleic Hydrazide 99.0+%, TCI America™

Catalog No. p-7131928
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M001525G 25 g
M0015500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M001525G Supplier TCI America Supplier No. M001525G

Chemical Identifiers

CAS 123-33-1
Molecular Formula C4H4N2O2
Molecular Weight (g/mol) 112.088
MDL Number MFCD00006665
InChI Key BGRDGMRNKXEXQD-UHFFFAOYSA-N
Synonym pyridazine-3,6-diol, maleic hydrazide, 3,6-dihydroxypyridazine, 3,6-pyridazinediol, stuntman, maleic acid hydrazide, vondalhyde, 3,6-pyridazinedione, 1,2-dihydro, 1,2-dihydro-3,6-pyridazinedione, antergon
PubChem CID 21954
ChEBI CHEBI:81771
IUPAC Name 1,2-dihydropyridazine-3,6-dione
SMILES C1=CC(=O)NNC1=O

Specifications

Color White to Yellow
pH 3.20 (50 g/L H2O soln, 25°C)
Quantity 25 g
Formula Weight 112.09
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Maleic Hydrazide
EINECSNumber (5)-0973
RTECSNumber UR5950000
TSCA Yes