missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid 99.0+%, TCI America™

Catalog No. M000625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M000625G 25 g
M0006500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M000625G Supplier TCI America Supplier No. M000625G

Chemical Identifiers

CAS 110-16-7
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00063177
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
Synonym maleic acid, cis-butenedioic acid, toxilic acid, maleinic acid, malenic acid, 2z-but-2-enedioic acid, 2-butenedioic acid z, maleate, cis-1,2-ethylenedicarboxylic acid, z-butenedioic acid
PubChem CID 444266
ChEBI CHEBI:18300
IUPAC Name (2E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

Melting Point 131°C
Color White
UN Number 3261
Formula Weight 116.07
Percent Purity ≥99.0% (T)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Maleic Acid
EINECSNumber (2)-1100
RTECSNumber OM9625000
TSCA Yes