missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid, NF, 99-101%, Spectrum™ Chemical

Catalog No. 18604148
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604148 12 kg Fiber Drum
18604146 25 g Amber Glass Bottle
18604147 100 g Amber Glass Bottle
18604149 500 g Amber Glass Bottle
18604145 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18604148 Supplier Spectrum Chemical Mfg Cor Supplier No. M120312KG
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1203, 110-16-7, C4H4O4

Spectrum™ Chemical Maleic Acid, NF

Chemical Identifiers

CAS 110-16-7
Molecular Formula C4H4O4
Molecular Weight (g/mol) 116.07
MDL Number MFCD00063177
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
IUPAC Name (2E)-but-2-enedioic acid
SMILES OC(=O)\C=C\C(O)=O

Specifications

CAS Max % 100%
Quantity 12 kg
Ignition Residue 0.1%
Packaging Fiber Drum
UN Number UN3261
Percent Purity 99 to 101%
Grade NF