missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maleic Acid, NF, 99-101%, Spectrum™ Chemical
Description
Spectrum™ Chemical Maleic Acid, NF
Specifications
Specifications
| CAS Max % | 100% |
| UN Number | UN3261 |
| Percent Purity | 99 to 101% |
| Grade | NF |
| Quantity | 50 kg |
| Ignition Residue | 0.1% |
| Packaging | Fiber Drum |