missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid Monoamide 98.0+%, TCI America™

Catalog No. M000325G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M000325G 25 g
M0003500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M000325G Supplier TCI America Supplier No. M000325G

Chemical Identifiers

CAS 557-24-4
Molecular Formula C4H5NO3
Molecular Weight (g/mol) 115.088
MDL Number MFCD00082354
InChI Key FSQQTNAZHBEJLS-UPHRSURJSA-N
Synonym Maleamic Acid
PubChem CID 5280451
ChEBI CHEBI:29045
IUPAC Name (Z)-4-amino-4-oxobut-2-enoic acid
SMILES C(=CC(=O)O)C(=O)N

Specifications

Melting Point 166°C
Color White
Quantity 25 g
Formula Weight 115.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Maleic Acid Monoamide