missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Maleic Acid, (cis-Butenedioic Acid), Reagents
Specifications
Specifications
| Melting Point | 130°C to 130.5°C |
| Boiling Point | 275°C |
| Color | Colorless/White |
| Grade | Reagent |
| Quantity | 1 kg |
| Packaging | Poly Bottle |
| Physical Form | Solid |
| Specific Gravity | 1.59 |
| Chemical Name or Material | Maleic Acid, (cis-Butenedioic Acid) |