missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid, 99%, Spectrum™ Chemical

Catalog No. p-7165825
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
100 g
25 g
250 g
5 kg
500 g
Packaging:
Amber Glass Bottle
Poly Pail
Catalog No. Quantity Packaging
18611467 25 g Amber Glass Bottle
18611466 100 g Amber Glass Bottle
18611468 250 g Amber Glass Bottle
18611471 500 g Amber Glass Bottle
18611465 1 kg Amber Glass Bottle
18611470 5 kg Poly Pail
6 options

Catalog No. 18611467

Supplier: Spectrum Chemical Mfg Cor M107525GM

Only null left

Chemical Identifiers

CAS 110-16-7

Specifications

CAS Max % 1
Percent Purity 99%
Grade Reagent
Quantity 25 g
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.