missing translation for 'onlineSavingsMsg'
Learn More

Maleic Acid, 99%, Spectrum™ Chemical

Catalog No. 18611470
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611470 5 kg Poly Pail
18611467 25 g Amber Glass Bottle
18611466 100 g Amber Glass Bottle
18611468 250 g Amber Glass Bottle
18611471 500 g Amber Glass Bottle
18611465 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611470 Supplier Spectrum Chemical Mfg Cor Supplier No. M10755KG
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 110-16-7

Specifications

CAS Max % 1
Quantity 5 kg
Packaging Poly Pail
Percent Purity 99%
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.