missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Male Rotating Luer Lock and Nut Assembly with 1/8 in. ID - CrystalVu - QC

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Nut and body are locked and rotate as a single unit. Precision engineered for smooth positive sealing. Color nut for easy recognition. This standard luer is ISO 80369-7 compliant. 

$197.59

Specifications

Certifications/Compliance Compliance Certificates Available
Description Class 7 Cleanroom Manufactured
Material CrystalVu (CVU)
For Use With (Application) Tubing connections; Medical device input/output ports; Drug and fluid transfer lines
Packaging Quantity 100 Pack
View More Specs

Products 4
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
01000275
View Documents Promotion Details
Berry Blue
Pack of 100 Each for $197.59
Only null left
 
01000276
View Documents Promotion Details
Lime Green
Pack of 100 Each for $197.59
Only null left
 
01000278
View Documents Promotion Details
Orange
Pack of 100 Each for $197.59
Only null left
 
01000280
View Documents Promotion Details
Violet
Pack of 100 Each for $197.59
Only null left
 
Description

Description

  • Single & Multi Use Applications
  • PVC-Free
  • ISO 80369-7 Compliant
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661, USP81, USP 88
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty and Services

Replacement if not satisfied. 12 Month return Policy.

Specifications

Specifications

Compliance Certificates Available
CrystalVu (CVU)
100 Pack
Rotating Luer Lock
Class 7 Cleanroom Manufactured
Tubing connections; Medical device input/output ports; Drug and fluid transfer lines
Out of direct sunlight, >20°F <100°F
Videos
SDS
Documents

Documents