missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Male Rotating Luer Lock and Nut Assembly with 1/4 in. ID - Medical Nylon - QC

Catalog No. 01000324
Change view
Click to view available options
Color:
Berry Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
01000324 Berry Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01000324
Only null left

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing. Nut and body are locked and rotate as a single unit. Precision engineered for smooth positive sealing. Color nut for easy recognition. This standard luer is ISO 80369-7 compliant. 

  • Single & Multi Use Applications
  • PVC-Free
  • ISO 80369-7 Compliant
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661, USP81, USP 88
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Compliance Certificates Available
Description Class 7 Cleanroom Manufactured
Material Nylon
For Use With (Application) Tubing connections; Medical device input/output ports; Drug and fluid transfer lines
Packaging Quantity 100 Pack
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Berry Blue
Product Type Rotating Luer Lock