Learn More
Iba Gmbh MagStrep Strep-TactinXT magnetic beads (5% suspensions), 2 ml
Catalog No. NC2604676
Encompass
MagStrep Strep-TactinXT beads can be used for the purification of strep-tagged fusion proteins in batch in single or high-throughput format.MagStrep Strep-TactinXT beads show low non-specific protein binding due to their agarose surface and high specificity of Strep-TactinXT towards Strep-tag. This results in outstanding target protein purity and makes the MagStrep Strep-TactinXT beads a perfect tool for the isolation of proteins or the identification of complexes via pull-downs, immunoprecipitation (IP), or co-immunoprecipitation (Co-IP). MagStrep Strep-TactinXT beads are suitable for fast small-scale purifications in reaction tubes as well as 96-well plates.
Catalog No. NC2604676 Supplier Iba Gmbh Supplier No. 25090002
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MagStrep Strep-TactinXT beads can be used for the purification of strep-tagged fusion proteins in batch in single or high-throughput format.MagStrep Strep-TactinXT beads show low non-specific protein binding due to their agarose surface and high specificity of Strep-TactinXT towards Strep-tag. This results in outstanding target protein purity and makes the MagStrep Strep-TactinXT beads a perfect tool for the isolation of proteins or the identification of complexes via pull-downs, immunoprecipitation (IP), or co-immunoprecipitation (Co-IP). MagStrep Strep-TactinXT beads are suitable for fast small-scale purifications in reaction tubes as well as 96-well plates.