missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAGPIX™ Calibration Kit

Catalog No. MPXCALK25
Change view
Click to view available options
Quantity:
25 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPXCALK25 25 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPXCALK25 Supplier Invitrogen™ Supplier No. LSMPXCALK25

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This convenient calibration kit contains xMAP™ calibrators for the MAGPIX™ system classification and reporter channels.

This convenient calibration kit contains xMAP™ calibrators for the MAGPIX™ system classification and reporter channels. Calibration normalizes the settings for the system and helps ensure optimal and consistent microsphere classification. The kit is intended to be used with the off-plate reagent block. Each kit provides 25 calibrations.

Each Kit includes:

  • Stripwells
  • A lot-specific CD for importing target values

The MAGPIX™ system is a versatile, compact, fluorescent detection system based on Luminex's proven xMAP™ technology. The system is a simple immunoassay platform designed for the simultaneous analysis of up to 50 intracellular and extracellular markers in single biological samples. The MAGPIX™ system provides users with a powerful multiplexing tool that offers higher sample throughput, increased assay flexibility, reduced sample volume, and lower costs than traditional ELISA and Western blotting.

Specifications

Product Line MAGPIX
Content And Storage • 25 strip wells (Cat. No. 13-52047)
• MAGPIX™ Calibrator Microspheres, 5 mL (MPXCAL-05)
• MAGPIX™ Calibration Kit CD (89-20233-00-001)

Store at 2°C to 8°C.
Quantity 25 Reactions
Shipping Condition Wet Ice
For Use With (Equipment) Flow Cytometer
Product Type Calibration Kit
How long does it take to acquire ProcartaPlex assay data on the different Luminex instruments?

With everything working smoothly, here is the approximate read time for one plate on different Luminex instruments:

- FLEXMAP 3D Systems and INTELLIFLEX Systems will read 96 well plates in ~20 min, and 384 well plates in ~75 min.
- Luminex 100/200 Systems will read 96 well plates in ~40 min.
- MAGPIX System will read 96 well plates in ~50-60 min. Please note that these are estimates for the read times of the plates only, and do not include time for calibration or set-up of the instrument.

For Research Use Only. Not for use in diagnostic procedures.