missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Mouse NK cell Enrichment Kit

Catalog No. 501125089
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125089 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125089 Supplier Invitrogen™ Supplier No. 8804682874

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Contains Sodium azide

Includes

  • MagniSort Mouse NK cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads A

Designed for the magnetic separation of mouse NK cells by negative selection. This kit will not isolate NKT cells. It has been optimized for the isolation of mouse NK cells from spleens utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave NK cells untouched and free in solution.

The MagniSort™ Mouse NK cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Mouse CD3 Biotin
Anti-Mouse CD4 Biotin
Anti-Mouse CD8 Biotin
Anti-Mouse CD19 Biotin
Anti-Mouse MHC Class II (I-A/I-E) Biotin
Anti-Mouse F4/80 Antigen Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse TER-119 Biotin

Specifications

Cell Type NK Cells
Product Type Cell Enrichment Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.