missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Mouse Hematopoietic Lineage Depletion Kit

Catalog No. 501125090
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125090 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125090 Supplier Invitrogen™ Supplier No. 8804682974
Contains Sodium azide

Includes

  • MagniSort Mouse Hematopoietic Lineage Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads B

Designed for the magnetic depletion of mouse hematopoietic cells with committed lineages from mouse bone marrow. It uses a cocktail of antibody specific for hematopoietic lineages and streptavidin-coated magnetic beads.

  • Lineage committed cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the lineage negative cells can be separated from lineage committed cells by decanting
  • After depletion, the enrichment of multi-potent progenitor cells can be verified by staining with Anti-Mouse CD34 (clone RAM34), and the enrichment of CD34- hematopoietic stem cell can be verified by additional staining with CD150 (clone mShad150), CD48 (clone HM48-1)
The MagniSortTM Mouse Hematopoietic Lineage Depletion Antibody Cocktail contains the following antibodies:
Anti-Mouse CD2 Biotin
Anti-Mouse CD3 Biotin
Anti-Mouse CD5 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD19 Biotin
Anti-Mouse CD45R Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse TER-119 Biotin

Specifications

Cell Type Hematopoietic Cells
Product Type Cell Depletion Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.