missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ MagniSort™ Mouse CD90.2 Positive Selection Kit

Catalog No. 501125074
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125074 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125074 Supplier Invitrogen™ Supplier No. 8802684474
Contains Sodium azide

Includes

  • MagniSort™ Anti-Mouse CD90.2 Biotin : 200 tests, 20μL/test
  • MagniSort™ Positive Selection Beads A

Designed for the magnetic separation of CD90.2+ cells by positive selection. It has been optimized for the isolation of CD90.2+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD90.2 antibody and streptavidin-coated magnetic beads.

  • CD90.2+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD90.2+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse CD90.2, clone 53-2.1
  • When the starting population of CD90.2 cells is less than 5% of the total cells, the amount of MagniSort™ Magnetic Positive Selection Beads to be used can be reduced to 10 μL/test to maximize purity

Specifications

Content And Storage 2° to 8°C. Do not freeze.
For Use With (Application) Magnetic Cell Separation
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 200 Tests