missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Mouse CD90.1 Positive Selection Kit

Catalog No. 501125073
Change view
Click to view available options
Quantity:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501125073 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 501125073 Supplier Invitrogen™ Supplier No. 8802684374
Contains Sodium azide

Includes

  • MagniSort™ Anti-Mouse CD90.1 Biotin : 200 tests, 20μL/test
  • MagniSort™ Positive Selection Beads A

Designed for the magnetic separation of CD90.1+ cells by positive selection. It has been optimized for the isolation of CD90.1+ cells from mouse spleens or lymph nodes utilizing a biotinylated Anti-Mouse CD90.1 antibody and streptavidin-coated magnetic beads.

  • CD90.1+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD90.1+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse/Rat CD90.1, clone HIS51
  • When the starting frequency of CD90.1 cells is less than 5% of the total cells, the amount of MagniSort™ Magnetic Positive Selection Beads to be used can be reduced to 10 μL/test to maximize purity

Specifications

Cell Type CD90.1+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Mouse

For Research Use Only. Not for use in diagnostic procedures.