missing translation for 'onlineSavingsMsg'
Learn More

eBioscience™ MagniSort™ Mouse CD8 Naïve T cell Enrichment Kit

Catalog No. 50728307
Change view
Click to view available options
Quantity:
200 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50728307 200 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Contains Sodium azide
Catalog No. 50728307 Supplier Thermo Fisher Scientific Supplier No. 8804682574
Contains Sodium azide

Includes

  • MagniSort Mouse CD8 Naïve T cell Enrichment Antibody Cocktail : 200 tests, 20μL/test
  • MagniSort Negative Selection Beads B

Designed for the magnetic separation of CD8 naïve T cells by negative selection. It has been optimized for the isolation of mouse CD8 naïve T cells from spleens or lymphnodes utilizing a biotinylated antibody cocktail and streptavidin-coated magnetic beads.

Undesired cells are bound by antibody and then magnetic beads that, when placed in a magnetic field, leave CD8 naïve T cells untouched and free in solution.

The MagniSort™ Mouse CD8 Naïve T cell Enrichment Antibody Cocktail contains the following antibodies:
Anti-Mouse CD4 Biotin
Anti-Mouse CD11b Biotin
Anti-Mouse CD19 Biotin
Anti-Mouse CD24 Biotin
Anti-Mouse CD44 Biotin
Anti-Mouse CD45R (B220) Biotin
Anti-Mouse CD49b (Integrin alpha 2) Biotin
Anti-Mouse Ly-6G (Gr-1) Biotin
Anti-Mouse γδ TCR Biotin
Anti-Mouse TER-119 Biotin

Specifications

Content And Storage 2° to 8°C. Do not freeze.
For Use With (Application) Magnetic Cell Separation
Format Aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Quantity 200 Tests